ENA308583874 Enamine

4-(aminomethyl)-2-ethoxyphenol hydrochloride

  • MDL number MFCD06797837
  • Cas Number : 105026-85-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68659

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68659
Availability details for
Quantity
  • Availability
  • Add to Cart