ENA308583884 Enamine

2-amino-N-methyl-8H-indeno[2,1-b]thiophene-3-carboxamide

  • MDL number MFCD11653115
  • Cas Number : 1094486-06-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95

Safety Information

msds https://www.enaminestore.com/catalog/EN300-68673
Availability details for
Quantity
  • Availability
  • Add to Cart