ENA308583893 Enamine

4-oxo-1-phenylcyclohexane-1-carbonitrile

  • MDL number MFCD00044816
  • Cas Number : 25115-74-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 110...114
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-21033

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-21033
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart