ENA308583905 Enamine

N-(prop-2-yn-1-yl)methanesulfonamide

  • MDL number MFCD14631470
  • Cas Number : 93501-84-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 56...58
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68707

Safety Information

msds https://www.enaminestore.com/catalog/EN300-68707
Availability details for
Quantity
  • Availability
  • Add to Cart