ENA308583908 Enamine

2-(4-methyl-1H-indol-3-yl)acetonitrile

  • MDL number MFCD09954796
  • Cas Number : 23084-32-4
  • Molecular Weight : 170.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-68711

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68711
Availability details for
Quantity
  • Availability
  • Add to Cart