ENA308583909 Enamine

6-oxo-N-(prop-2-yn-1-yl)-1,6-dihydropyridine-3-carboxamide

  • MDL number MFCD11174677
  • Cas Number : 1040070-36-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68712

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68712
Availability details for
Quantity
  • Availability
  • Add to Cart