ENA308583911 Enamine

3-[(4-amino-3-methylphenoxy)methyl]-4-fluorobenzonitrile

  • MDL number MFCD09811896
  • Cas Number : 1016509-76-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-68714

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68714
Availability details for
Quantity
  • Availability
  • Add to Cart