ENA308583912 Enamine

2-(4-methylphenyl)propanenitrile

  • MDL number MFCD11036541
  • Cas Number : 75920-45-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68715

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68715
Availability details for
Quantity
  • Availability
  • Add to Cart