ENA308583928 Enamine

3-[(2-nitrophenyl)amino]propanenitrile

  • MDL number MFCD11043170
  • Cas Number : 33141-24-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68739

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68739
Availability details for
Quantity
  • Availability
  • Add to Cart