ENA308583932 Enamine

2-methylpyrimidin-5-amine

  • MDL number MFCD09608109
  • Cas Number : 39889-94-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 155-159
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68746

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68746
Availability details for
Quantity
  • Availability
  • Add to Cart