ENA308583943 Enamine

3-[(3-amino-4-fluorophenyl)formamido]propanamide

  • MDL number MFCD12136317
  • Cas Number : 1157000-92-4
  • Molecular Weight : 225.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-68759

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68759
Availability details for
Quantity
  • Availability
  • Add to Cart