ENA308583945 Enamine

3-ethoxy-4-(2-methoxy-2-oxoethoxy)benzoic acid

  • MDL number MFCD09938653
  • Cas Number : 1016829-67-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-68761

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68761
Availability details for
Quantity
  • Availability
  • Add to Cart