ENA308583965 Enamine

2-(2-chloro-6-fluorophenyl)acetamide

  • MDL number MFCD00120063
  • Cas Number : 895903-54-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68799

Safety Information

msds https://www.enaminestore.com/catalog/EN300-68799
Availability details for
Quantity
  • Availability
  • Add to Cart