ENA308583969 Enamine

2-amino-6-fluorobenzene-1-sulfonamide

  • MDL number MFCD12913294
  • Cas Number : 1161930-99-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-68803

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68803
Availability details for
Quantity
  • Availability
  • Add to Cart