ENA308583972 Enamine

1-(4-bromophenyl)pyrrolidin-3-amine

  • MDL number MFCD11894490
  • Cas Number : 1181332-98-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-68807
Availability details for
Quantity
  • Availability
  • Add to Cart