ENA308583975 Enamine

1-[(piperidin-3-yl)methyl]azepane

  • MDL number MFCD05215126
  • Cas Number : 896053-57-9
  • Molecular Weight : 196.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68810

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68810
Availability details for
Quantity
  • Availability
  • Add to Cart