ENA308583976 Enamine

2-(4-fluorophenyl)-2-methylpropanoic acid

  • MDL number MFCD11036916
  • Cas Number : 93748-19-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 89...91
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68812

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-68812
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart