ENA308583985 Enamine

2-(hydroxymethyl)benzoic acid

  • MDL number MFCD02151998
  • Cas Number : 612-20-4
  • Molecular Weight : 152.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 120-122
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-68822

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68822
Availability details for
Quantity
  • Availability
  • Add to Cart