ENA308583990 Enamine

N-propyl-1,2,3,4-tetrahydroquinoline-6-sulfonamide

  • MDL number MFCD11189656
  • Cas Number : 952931-09-8
  • Molecular Weight : 254.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-68827

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68827
Availability details for
Quantity
  • Availability
  • Add to Cart