ENA308583995 Enamine

2-(4-benzyl-2,5-dioxoimidazolidin-1-yl)-3-phenylpropanoic acid

  • MDL number MFCD17167211
  • Cas Number : 13589-09-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68835

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68835
Availability details for
Quantity
  • Availability
  • Add to Cart