ENA308583996 Enamine

2-(4-chlorophenyl)-4-fluoroaniline

  • MDL number MFCD12824033
  • Cas Number : 188731-34-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-68839

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68839
Availability details for
Quantity
  • Availability
  • Add to Cart