ENA308584000 Enamine

2-[2-(1H-1,2,4-triazol-1-yl)ethoxy]ethan-1-ol

  • MDL number MFCD12785460
  • Cas Number : 1248039-33-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68844

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68844
Availability details for
Quantity
  • Availability
  • Add to Cart