ENA308584002 Enamine

2-(1,3-dioxaindan-5-yl)-1,3-thiazole-5-carboxylic acid

  • MDL number MFCD07376772
  • Cas Number : 1094385-70-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68846

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68846
Availability details for
Quantity
  • Availability
  • Add to Cart