ENA308584003 Enamine

5-[3-(trifluoromethyl)phenyl]-1,3,4-oxadiazol-2-amine

  • MDL number MFCD01710639
  • Cas Number : 5711-64-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68847

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68847
Availability details for
Quantity
  • Availability
  • Add to Cart