ENA308584005 Enamine

4-[5-(pyridin-3-yl)-1,2,4-oxadiazol-3-yl]phenol

  • MDL number MFCD10693672
  • Cas Number : 40678-80-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68849

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68849
Availability details for
Quantity
  • Availability
  • Add to Cart