ENA308584017 Enamine

2-(chloromethyl)-1-ethyl-4-fluoro-1H-1,3-benzodiazole

  • MDL number MFCD12149781
  • Cas Number : 1183366-81-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 115...117
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68863

Safety Information

msds https://www.enaminestore.com/catalog/EN300-68863
Availability details for
Quantity
  • Availability
  • Add to Cart