ENA308584019 Enamine

[(3-methylphenyl)methyl][(pyridin-2-yl)methyl]amine dihydrochloride

  • MDL number MFCD17167216
  • Cas Number : 1252441-99-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68865

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68865
Availability details for
Quantity
  • Availability
  • Add to Cart