ENA308584020 Enamine

1-{4-[(4-fluorophenyl)methoxy]phenyl}ethan-1-amine

  • MDL number MFCD09730929
  • Cas Number : 953901-43-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 52...54
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68866

Safety Information

msds https://www.enaminestore.com/catalog/EN300-68866
Availability details for
Quantity
  • Availability
  • Add to Cart