ENA308584024 Enamine

2-amino-4-methoxy-N-methylbenzene-1-sulfonamide

  • MDL number MFCD11118285
  • Cas Number : 1036570-75-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68876

Safety Information

msds https://www.enaminestore.com/catalog/EN300-68876
Availability details for
Quantity
  • Availability
  • Add to Cart