ENA308584027 Enamine

4-[3-(aminomethyl)pentan-3-yl]-1,2-dichlorobenzene

  • MDL number MFCD12816650
  • Cas Number : 1183596-21-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68879

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68879
Availability details for
Quantity
  • Availability
  • Add to Cart