ENA308584029 Enamine

2-methyl-2-(4-methylphenyl)propanoic acid

  • MDL number MFCD11036959
  • Cas Number : 20430-18-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 78-80
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-68881

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68881
Availability details for
Quantity
  • Availability
  • Add to Cart