ENA308584037 Enamine

1-(3-chloro-4-methoxyphenyl)ethan-1-ol

  • MDL number MFCD12786788
  • Cas Number : 124829-13-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68892

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68892
Availability details for
Quantity
  • Availability
  • Add to Cart