ENA308584039 Enamine

[(2-methylphenyl)methyl][(pyridin-2-yl)methyl]amine dihydrochloride

  • MDL number MFCD17167219
  • Cas Number : 1252334-11-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Product Link https://www.enaminestore.com/catalog/EN300-68894

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68894
Availability details for
Quantity
  • Availability
  • Add to Cart