ENA308584041 Enamine

tert-butyl N-{[methyl(pyridin-2-ylmethyl)carbamoyl]methyl}carbamate

  • MDL number MFCD17167221
  • Cas Number : 1258649-76-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68896

Safety Information

msds https://www.enaminestore.com/catalog/EN300-68896
Availability details for
Quantity
  • Availability
  • Add to Cart