ENA308584042 Enamine

N-(2-methylcyclohexyl)-2-(piperazin-1-yl)acetamide

  • MDL number MFCD11183513
  • Cas Number : 79379-27-4
  • Molecular Weight : 239.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name N-(2-methylcyclohexyl)-2-(1-piperazinyl)acetamide
Inchi Code 1S/C13H25N3O/c1-11-4-2-3-5-12(11)15-13(17)10-16-8-6-14-7-9-16/h11-12,14H,2-10H2,1H3,(H,15,17)
Inchi Key VADNNDPBXVAQTI-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-68897

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68897
Availability details for
Quantity
  • Availability
  • Add to Cart