ENA308584043 Enamine

2-(2-methylphenyl)acetamide

  • MDL number MFCD08589450
  • Cas Number : 40089-14-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-68898

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68898
Availability details for
Quantity
  • Availability
  • Add to Cart