ENA308584046 Enamine

N-cyclohexyl-N-methyl-2-(piperazin-1-yl)acetamide

  • MDL number MFCD11183506
  • Cas Number : 1040324-59-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68906

Safety Information

msds https://www.enaminestore.com/catalog/EN300-68906
Availability details for
Quantity
  • Availability
  • Add to Cart