ENA308584049 Enamine

N-[1-(hydrazinecarbonyl)ethyl]methanesulfonamide

  • MDL number MFCD09836487
  • Cas Number : 117267-37-5
  • Molecular Weight : 181.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68909

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68909
Availability details for
Quantity
  • Availability
  • Add to Cart