ENA308584050 Enamine

3-fluoro-4-[(3-fluorophenyl)methoxy]aniline

  • MDL number MFCD11621723
  • Cas Number : 231278-65-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-68910

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68910
Availability details for
Quantity
  • Availability
  • Add to Cart