ENA308584052 Enamine

2-[3-(trifluoromethyl)phenyl]acetamide

  • MDL number MFCD11643205
  • Cas Number : 22902-93-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68912

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68912
Availability details for
Quantity
  • Availability
  • Add to Cart