ENA308584055 Enamine

3-(4-cyanophenoxy)benzoic acid

  • MDL number MFCD00447014
  • Cas Number : 313549-93-8
  • Molecular Weight : 239.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-68915

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68915
Availability details for
Quantity
  • Availability
  • Add to Cart