ENA308584062 Enamine

N-(4,5-dihydro-1,3-thiazol-2-yl)pyridin-3-amine dihydrochloride

  • MDL number MFCD17167225
  • Cas Number : 1252444-09-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68922

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68922
Availability details for
Quantity
  • Availability
  • Add to Cart