ENA308584064 Enamine

1,3-dichloro-2-isothiocyanato-4-methylbenzene

  • MDL number MFCD11117350
  • Cas Number : 1036587-85-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68924

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68924
Availability details for
Quantity
  • Availability
  • Add to Cart