ENA308584068 Enamine

N-butyl-2-chloro-N-ethylacetamide

  • MDL number MFCD03001141
  • Cas Number : 2567-58-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-23006

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-23006
Availability details for
Quantity
  • Availability
  • Add to Cart