ENA308584069 Enamine

4-[2-(2-aminophenoxy)ethyl]-1,2,3,4-tetrahydroquinoxalin-2-one

  • MDL number MFCD13673375
  • Cas Number : 1099658-11-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68929

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68929
Availability details for
Quantity
  • Availability
  • Add to Cart