ENA308584072 Enamine

4-chloro-2-[(oxan-4-yl)amino]benzonitrile

  • MDL number MFCD12118927
  • Cas Number : 939816-62-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68932

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68932
Availability details for
Quantity
  • Availability
  • Add to Cart