ENA308584074 Enamine

4-chloro-2-(cyclopentylamino)benzonitrile

  • MDL number MFCD12118942
  • Cas Number : 939812-57-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68934

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68934
Availability details for
Quantity
  • Availability
  • Add to Cart