ENA308584076 Enamine

ethyl 5-(chlorosulfonyl)-2-fluorobenzoate

  • MDL number MFCD12169093
  • Cas Number : 1155985-73-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68936

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68936
Availability details for
Quantity
  • Availability
  • Add to Cart