ENA308584079 Enamine

2-(1H-imidazol-2-yl)-1-(4-methylphenyl)ethan-1-one hydrochloride

  • MDL number MFCD17167230
  • Cas Number : 52855-77-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-68941

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68941
Availability details for
Quantity
  • Availability
  • Add to Cart