ENA308584089 Enamine

2-(1H-imidazol-2-yl)-1-(2-methylphenyl)ethan-1-one hydrochloride

  • MDL number MFCD17167231
  • Cas Number : 1252206-09-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68952

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68952
Availability details for
Quantity
  • Availability
  • Add to Cart